▸
Salts and Inorganics
▸
Inorganic Salts
▸
Alkali Metal Salts
▸
Sodium Phosphate Dibasic Anhydrous (Granular or Powder/Certified ACS), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Sodium Phosphate Dibasic Anhydrous (Granular or Powder/Certified ACS), Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
USFS374500
500 g
Glass Bottle
Catalog number
USFS374500
Price (SGD)
62.00
Each
Add to cart
Quantity:
500 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (SGD)
62.00
Each
Add to cart
Specifications
Chemical Name or Material
Sodium Phosphate Dibasic Anhydrous
CAS
7558-79-4
ChemAlert Storage Symbol
Gray
Color
White
Health Hazard 1
Signal Word: CAUTION!
View more