▸
Organic compounds
▸
Organooxygen compounds
▸
Alcohols and polyols
▸
Propylene Glycol (USP/FCC), Fisher Chemical™
Element Search
Structure Search
Propylene Glycol (USP/FCC), Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
USFP35520
20 L
Lined Steel Drum
Catalog number
USFP35520
Price (SGD)
-
Quantity:
20 L
Packaging:
Lined Steel Drum
Request bulk or custom format
Specifications
Chemical Name or Material
Propylene Glycol
Acidity
Pass Test (FCC/USP)
CAS
57-55-6
ChemAlert Storage Symbol
Gray
Health Hazard 2
WARNING!
View more
Propylene Glycol (USP/FCC)