Methylene Chloride, Optima™ for HPLC and GC, Fisher Chemical™
1LT Dichloromethane, for residue analysis, Distol™, stabilised with amylene
Have Questions?
Catalog Number
Quantity
Packaging
USFD1514
4 L
Amber Soda Lime Glass
Catalog number USFD1514
Price (SGD)
163.00
Bottle
Quantity:
4 L
Packaging:
Amber Soda Lime Glass
Request bulk or custom format
Price (SGD)
163.00
Bottle
Specifications
Chemical Name or MaterialMethylene Chloride
Cleanup SymbolSolvent
Melting Point-97°C
Absorbance1 AU max. at 233nm, 0.12 AU max. at 240nm, 0.005 AU max. at 254nm
CAS75-09-2
View more
Screened for fluorescing and UV-absorbing contaminants to reduce interferences for smoother baselines
Impurity profile is also reported using gas chromatography and three different detectors (ECD, FID, and MS) to demonstrate suitable for different analyses
Contains approx. 15- 200 ppm amylene as preservative