▸
Solvents
▸
Ethanol
▸
Alcohol (HPLC, Meets ACS), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Alcohol (HPLC, Meets ACS), Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
USFA9954
4 L
Glass Bottle
Catalog number
USFA9954
Price (SGD)
84.00
Bottle
Add to cart
Quantity:
4 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (SGD)
84.00
Bottle
Add to cart
Specifications
Chemical Name or Material
Alcohol
Cleanup Symbol
Solvent
Melting Point
-114.1°C
Absorbance
0.04 AU max. at 254nm, 0.35 AU max. at 220nm, 0.65 AU max. at 210nm, 1 AU max. at 205nm
CAS
64-17-5
View more