Acetone, Optima™, for HPLC and GC, Fisher Chemical™
Acetone, Optima™, for HPLC and GC, Fisher Chemical™
Acetone, Optima™, for HPLC and GC, Fisher Chemical™
Acetone, Optima™, for HPLC and GC, Fisher Chemical™
Fisher Chemical
Acetone, Optima™, for HPLC and GC, Fisher Chemical™
High purity and versatile solvent suitable for HPLC, spectrophotometry, and pesticide residue analysis
Have Questions?
Change viewbuttonViewtableView
Quantity:
4 L
1 L
Packaging:
View all
Amber Soda Lime Glass
Catalog number USFA9294
Price (SGD)
158.00
Bottle
Quantity:
4 L
Packaging:
Amber Soda Lime Glass
Request bulk or custom format
Price (SGD)
158.00
Bottle
Specifications
Chemical Name or MaterialAcetone
pH7
Cleanup SymbolSolvent
Melting Point-95°C
Absorbance1.00 AU max. 330nm, 0.06 AU max. at 340nm, 0.010 AU max. 350nm, 0.01 AU max. (350 to 400nm)
View more
  • Screened for fluorescing contaminants, and UV-absorbing contaminants in the 300-400nm range, to reduce interferences for smoother baselines
  • Tested to ensure minimum halogenated residue levels (≤10 ng/L) to demonstrate suitability for pesticide analysis
  • Also meets ACS specifications
  • Intended for research use only