Check Order Status
Ammonium Hydroxide (Certified ACS Plus), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Ammonium Hydroxide (Certified ACS Plus), Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
USFA669500
500 mL
Glass Bottle
Catalog number
USFA669500
Price (SGD)
75.00
EA
Add to cart
Quantity:
500 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (SGD)
75.00
EA
Add to cart
Specifications
Chemical Name or Material
Ammonium Hydroxide
pH
12
Cleanup Symbol
Caustic
Color
Colorless
Name Note
about 14.8N
View more