Check Order Status
Ammonium Chloride (Crystalline/Certified ACS), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Ammonium Chloride (Crystalline/Certified ACS), Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
USFA661500
500 g
Poly Bottle
Catalog number
USFA661500
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Poly Bottle
Request bulk or custom format
Specifications
Boiling Point
520°C
Chemical Name or Material
Ammonium Chloride
Melting Point
340°C
CAS
12125-02-9
ChemAlert Storage Symbol
Gray
View more