Check Order Status
Hydrochloric Acid (NF/FCC), Fisher Chemical
Element Search
Structure Search
Fisher Chemical
Hydrochloric Acid (NF/FCC), Fisher Chemical
Have Questions?
Catalog Number
Quantity
Packaging
USFA481212
2.5 L
Glass Bottle
Catalog number
USFA481212
Price (SGD)
119.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (SGD)
119.00
EA
Add to cart
Specifications
Chemical Name or Material
Hydrochloric Acid
pH
1
Cleanup Symbol
Acid
Color
Colorless
CAS
7647-01-0
View more