▸
Solvents
▸
Isopropanols
▸
2-Propanol, ACS Grade, Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
2-Propanol, ACS Grade, Fisher Chemical™
In addition to meeting or exceeding the latest ACS specifications, are analyzed for more than 16 metals; Actual lot of analysis on label
Have Questions?
Change view
Quantity:
200 L
Packaging:
Steel Drum
Catalog number
USFA416200
Price (SGD)
3,511.00
Each
Add to cart
Quantity:
200 L
Packaging:
Steel Drum
Request bulk or custom format
Price (SGD)
3,511.00
Each
Add to cart
Specifications
Chemical Name or Material
2-Propanol
pH
7
Cleanup Symbol
Solvent
Melting Point
-89.5°C
CAS
67-63-0
View more