Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Chemicals
▸
Solvents
▸
Ethanol
▸
Ethanol, Anhydrous (Histological), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Ethanol, Anhydrous (Histological), Fisher Chemical™
Used in histology, pathology, and cell biology applications
Have Questions?
Change view
Quantity:
4 L
20 L
Packaging:
View all
Poly Bottle
Catalog number
USFA405P4
Price (SGD)
408.00
Each
Add to cart
Quantity:
4 L
Packaging:
Poly Bottle
Request bulk or custom format
Price (SGD)
408.00
Each
Add to cart
Specifications
Chemical Name or Material
Ethanol, Anhydrous
Melting Point
-90°C
CAS
64-17-5
ChemAlert Storage Symbol
Red
CAS
108-10-1
View more