Check Order Status
Sulfuric Acid (ACS Grade), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Sulfuric Acid (ACS Grade), Fisher Chemical™
Approx. Normality is 36
Have Questions?
Change view
Quantity:
2.5 L
500 mL
Packaging:
View all
PPQ Resin Bottle
Catalog number
USFA300C212
Price (SGD)
665.00
CS
Add to cart
Quantity:
2.5 L
Packaging:
PPQ Resin Bottle
Request bulk or custom format
Price (SGD)
665.00
CS
Add to cart
Specifications
Chemical Name or Material
Sulfuric Acid
pH
0.3
Cleanup Symbol
Acid
Color
Colorless to Brown
CAS
7664-93-9
View more