Formic Acid, 99.0+%, Optima™ LC/MS Grade, Fisher Chemical™
Formic Acid, 99.0+%, Optima™ LC/MS Grade, Fisher Chemical™
Formic Acid, 99.0+%, Optima™ LC/MS Grade, Fisher Chemical™
Fisher Chemical
Formic Acid, 99.0+%, Optima™ LC/MS Grade, Fisher Chemical™
Suitable as a mobile phase additive for LC/MS and UHPLC/MS
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
10 x 1 mL
Packaging:
View all
Nalgene™ Poly Bottle
Catalog number USFA11750
Price (SGD)
140.00
Each
Quantity:
50 mL
Packaging:
Nalgene™ Poly Bottle
Request bulk or custom format
Price (SGD)
140.00
Each
Specifications
Absorbance0.01 AU max. at 254nm, 0.55 AU max. at 230nm, 0.85 AU max. at 220nm, 1.25 AU max. at 210nm
Chemical Name or MaterialFormic Acid
Cleanup SymbolAcid
pH2.1
CAS64-18-6
View more