-80°C Benchtop Freezer
Actual product may vary
-80°C Benchtop Freezer
-80°C Benchtop Freezer
-80°C Benchtop Freezer
Protect precious samples with this bentchop freezer, ideal for personal or clinical trial applications.
Have Questions?
Catalog NumberElectrical RequirementsPlug Type
ULT185-5-V230 V 50 HzP7
Catalog number ULT185-5-V
HSN 84183090
Price (SGD)
-
Electrical Requirements:
230 V 50 Hz
Plug Type:
P7
Protect your precious samples with the Thermo Scientific -80°C Benchtop Freezer, which is ideal for personal or clinical trial applications offering a full 1 cu. ft. (28L) capacity.
Cabinet Construction:
  • Compact 25.2 x 30.7 in. (640 mm x 781 mm) footprint
  • Industrial-grade hermetically sealed refrigeration compressors
  • All-steel cabinet with high-impact epoxy finish for easy cleaning
  • 4 in. (102 mm) foamed-in-place, CFC-free urethane insulation to protect product against high ambient conditions
  • Heavy-duty counterbalanced lid with key lock
  • Thermal interior sublid
  • Air-cooled refrigeration systems to maintain design temperatures in warm 32°C (90°F) ambient conditions
Control System:
  • Solid-state control system with adjustable temperature setpoint
  • Digital temperature display resolution to 1°C
  • Push button setpoint display
  • Key operated main power switch
Alarm System:
  • Integrated preset overtemperature safety alarm with battery back-up and audible warning of temperature deviation
  • Exterior alarm contacts for connection to remote monitoring system
Specifications
DescriptionULT Freezer, 1CF(28L), 230V/50Hz - EU Plug
TypeUltra Low Freezer
Capacity (English)1 cu. ft.
Capacity (Metric)28 L
Temperature Range-50°C to -80°C
Cabinet MaterialSteel
InsulationWater-Blown Polyurethane Foam (SNAP-compliant)
InteriorPainted Steel
Exterior FinishEpoxy Coated Steel
Certifications/ComplianceCE
StyleBenchtop
DisplayPush Button LED
Product LineThermo Scientific
Water CooledNot Available
Dimensions (D x W x H) Interior12 x 15.3 x 9.6 in. (30.5 x 38.7 x 24.5 cm)
Dimensions (L x W x H) Exterior25.5 x 30.75 x 20 in. (64.8 x 78.1 x 50.8 cm)
Footprint (English)0.106 cu. ft.
Footprint (Metric)0.03 cu. meters
Monitoring OptionsAlarm, Recorder
Access SecurityKey Lock
Chart Recorder7-day Inkless (Optional)
Data OutputsNone
On-Board DataloggingNot Available
Operating ModesStandard Operation
Setpoint SecurityNot Available
Backup SystemsNot Available
Amperage12 A
Plug TypeP7
Voltage230 V
Electrical Requirements230 V 50 Hz
Frequency50 Hz
Line Voltage IndicatorNone
Vial Capacity1,600 x 2mL
CompressorHermetic
Shipping Weight (English)190 lb.
Shipping Weight (Metric)86 kg
HoldsBoxes (16)
Unit SizeEach