Check Order Status
Ethanol Absolute 99.8+%, Certified AR for Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™
Element Search
Structure Search
Ethanol Absolute 99.8+%, Certified AR for Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™
C2H6O, CAS Number-64-17-5, 2.5L, 78 deg.C, CHEBI:16236, Colorless, 46.07g/mol, LFQSCWFLJHTTHZ-UHFFFAOYSA-N, ethanol, -114 deg.C, 3568, 46.069, Amber glass bottle, Liquid, 702, CCO | CAS: 64-17-5 | C2H6O | 46.069 g/mol
Have Questions?
Change view
Quantity:
2.5 L
1 L
Packaging:
HDPE plastic bottle
Amber glass bottle
Catalog number
UKFE0650DFC17
also known as UKFE/0650DF/C17
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Packaging:
HDPE plastic bottle
Request bulk or custom format
Specifications
Packaging
HDPE plastic bottle
CAS
64-17-5
Physical Form
Liquid
Molecular Formula
C2H6O
MDL Number
3568
View more
C2H6O, CAS Number-64-17-5, 2.5L, 78 deg.C, CHEBI:16236, Colorless, 46.07g/mol, LFQSCWFLJHTTHZ-UHFFFAOYSA-N, ethanol, -114 deg.C, 3568, 46.069, Amber glass bottle, Liquid, 702, CCO