Dimethyl Sulfoxide, Certified AR for Analysis, Fisher Chemical™
Dimethyl Sulfoxide, Certified AR for Analysis, Fisher Chemical™
Dimethyl Sulfoxide, Certified AR for Analysis, Fisher Chemical™
Dimethyl Sulfoxide, Certified AR for Analysis, Fisher Chemical™
Dimethyl Sulfoxide, Certified AR for Analysis, Fisher Chemical™
C2H6OS, CAS Number-67-68-5, demasorb, demsodrox, methyl sulfoxide, dimexide, dimethyl sulfoxide, dimethylsulfoxide, demavet, methane, sulfinylbis, dimethyl sulphoxide, dmso, 2.5L, 189 deg.C, CHEBI:28262, Colorless, 78.13g/mol, IAZDPXIOMUYVGZ-UHFFFAOYSA-N, methylsulfinylmethane, 18.4 deg.C, 2089 | CAS: 67-68-5 | C2H6OS | 78.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
500 mL
Packaging:
HDPE Bottle
Catalog number UKFD4121PB17
also known as UKFD/4121/PB17
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Packaging:
HDPE Bottle
Request bulk or custom format
Specifications
Melting Point18.4°C
CAS67-68-5
ColorColorless
Boiling Point189°C
Quantity2.5 L
View more
C2H6OS, CAS Number-67-68-5, demasorb, demsodrox, methyl sulfoxide, dimexide, dimethyl sulfoxide, dimethylsulfoxide, demavet, methane, sulfinylbis, dimethyl sulphoxide, dmso, 2.5L, 189 deg.C, CHEBI:28262, Colorless, 78.13g/mol, IAZDPXIOMUYVGZ-UHFFFAOYSA-N, methylsulfinylmethane, 18.4 deg.C, 2089