Check Order Status
Acetone, for HPLC
Element Search
Structure Search
Acetone, for HPLC
C3H6O, CAS Number-67-64-1, 2.5L, 56 deg.C, Colorless, 58.08g/mol, -95 deg.C, 8765, Amber glass bottle, Liquid, 247mbar at 20 deg.C, 0.32 mPaS at 20 deg.C
Have Questions?
Catalog Number
Quantity
UKFA060617
also known as UKFA/0606/17
2.5 L
Catalog number
UKFA060617
also known as UKFA/0606/17
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Request bulk or custom format
Specifications
pH
7
Melting Point
-95°C
CAS
67-64-1
Color
Colorless
Boiling Point
56°C
View more
C3H6O, CAS Number-67-64-1, 2.5L, 56 deg.C, Colorless, 58.08g/mol, -95 deg.C, 8765, Amber glass bottle, Liquid, 247mbar at 20 deg.C, 0.32 mPaS at 20 deg.C