TSX Universal Series General Purpose Ultra-Low Freezer
Actual product may vary
TSX Universal Series General Purpose Ultra-Low Freezer
TSX Universal Series General Purpose Ultra-Low Freezer
TSX Universal Series General Purpose Ultra-Low Freezer
TSX Universal Series General Purpose Ultra-Low Freezer
TSX Universal Series General Purpose Ultra-Low Freezer
TSX Universal Series General Purpose Ultra-Low Freezer
TSX Universal Series General Purpose Ultra-Low Freezer
TSX Universal Series General Purpose Ultra-Low Freezer
TSX Universal Series General Purpose Ultra-Low Freezer
TSX Universal Series General Purpose Ultra-Low Freezer
TSX Universal Series Ultra-Low Freezers - The next generation of our most trusted ULT platform adapts to your needs, reduces your environmental impact, and is built on our most robust design ever.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberModelHoldsRack CapacityCapacity (Metric)
TSX60086FAGeneral Purpose Ultra-Low Freezer 6002 in. Boxes (600)24 Racks816 L
TSX40086FAGeneral Purpose Ultra-Low Freezer 4002 in. Boxes (400)16 Racks549 L
TSX50086FAGeneral Purpose Ultra-Low Freezer 5002 in. Boxes (500)20 Racks682 L
TSX70086FAGeneral Purpose Ultra-Low Freezer 7002 in. Boxes (700)28 Racks949 L
TSX70086FA-ULTSUltra-Low Freezer 7002 in. boxes (700)28 Racks950 L
Catalog number TSX60086FA
HSN 84184090
Price (SGD)
28,195.00
Each
Model:
General Purpose Ultra-Low Freezer 600
Holds:
2 in. Boxes (600)
Rack Capacity:
24 Racks
Capacity (Metric):
816 L
Price (SGD)
28,195.00
Each
TSX Universal—Over 80 years of innovation packed into the next-gen ultra-low freezer. Recognizing the impact of your work, we prioritize sample security and reliability, and our legacy of crafting state-of-the-art lab equipment means our focus remains on user-centric design. The TSX Universal heralds a new era in performance with Universal V-Drive technology, delivering tighter control and faster recovery times. Coupled with an expanded setpoint range, universal voltage, whisper-quiet operation, simplified software updates and much more, TSX Universal seamlessly adapts to your workflow. Redefining sustainability, it is ENERGY STAR certified with a 32% efficiency improvement, ACT labeled, and is produced in our Zero-Waste to Landfill certified facility. TSX Universal ULT—The pinnacle of reliability, sustainability, and innovation.
Reliability at its core:
  • Manufactured excellence: built to the highest standards at our ISO 13485:2016 facilities
  • Proven durability: 10k+ hours of intensive lab and field runtime testing to help ensure long-term reliability
  • Proven quality: each unit undergoes rigorous end-of-line testing for quality control
  • Complete transparency: shipped with factory documentation and a certification of calibration*
  • Peace of mind: backed by our 12-year warranty** for long-term confidence
Built to perform, designed to protect:
  • Enhanced performance with universal V-Drive technology: adapts to user activity to help maintain peak performance and protect what matters most—your samples.
  • Stable temperatures: less than 4°C variation helps keep samples safe anywhere in the cabinet
  • Fast recovery: industry-leading 15-minute door recovery helps minimize temperature fluctuations and downtime
  • Rapid cooling, faster setup: reaches -80°C in under 5 hours—41% faster than previous models
  • Cost effective: uses 33% less energy than previous models , helping reduce costs and environmental impact
  • Quick sample access with active pressure relief: innovative pressure relief technology enables re-entry in just 30 seconds so you can retrieve samples as frequently as needed, helping minimize disruptions
  • Maximum protection & compact footprint: advanced vacuum insulation and environmentally friendly 0 GWP, water-blown foam insulation safeguard samples while providing a compact footprint
Greener by design
  • Less hazardous: uses non-hydrofluorocarbon (HFC) refrigerants and foam insulation which has a 0 GWP and Ozone Depleting Potential (ODP) and is water-blown, helping reduce chemical emissions and outgassing
  • Less waste: contains up to 70% recycled materials, with 90% of waste diverted from landfills at our zero-waste facility***†
  • More energy efficient: ENERGY STAR® certified†, helping reduce lab energy costs and environmental footprint
  • Responsibly packaged: uses non-hydrofluorocarbon (non-HFC) refrigerants with low Global Warming Potential (GWP) and foam insulation, with 0 GWP and Ozone Depleting Potential (ODP), that is water-blown, helping reduce chemical emissions and outgassing
  • Extended life: offers a 12-year warranty, longer than any comparable product on the market
Enhanced experience, optimized for ease
  • Lab-ready convenience: universal voltage, multiple size options, and whisper-quiet operation for easy installation and compatibility
  • Reliable storage and organization: ergonomic handle helps ensure an airtight seal to prevent temperature fluctuations, while adjustable racking maximizes storage efficiency
  • Enhanced security and monitoring: 7 in. UI helps streamline operations, optional HID access enhances security, built-in power and outputs enable real-time monitoring and diagnostics, and an optional lightbar allows instant unit status visibility
  • Easy frost management: 7-point gasket design with an adjustable perimeter heater helps minimize frost build up for easier maintenance
  • Seamless integration: built-in ports for LN2, CO2, and monitoring probes helps ensure compatibility with your lab setup
*Shipped documents:
Certificate of conformance, CE declaration of conformance, certificate of calibration, QC test log sheets, and QC temperature graphs.
**Restrictions and exclusions apply. Warranty is void if the unit is not maintained as set forth in the operation.
***Zero waste defined as 90% of non-hazardous waste diverted from landfill, waste to energy and incineration.
† Information not applicable to TSX Universal ULT freezers with '-ULTS' in the part number (TSX40086FA-ULTS, TSX50086FA-ULTS, TSX60086FA-ULTS.)
Specifications
DescriptionTSX Universal Series Ultra-Low Freezers, 600-Box Capacity, 100/230 V, 50/60 Hz
TypeUltra-Low Freezer, Upright
Capacity (English)28.8 cu. ft.
Capacity (Metric)816 L
Temperature Range-40°C to -86°C
Temperature Resolution0.1°C
Temperature Stability0.2°C
Door StyleOuter (1), Inner (2)
No. of Doors1
Door Opening Recovery17 min. (20°C Ambient Running at -75°C)
Cabinet MaterialScratch-resistant Painted Steel
Chamber MaterialCold-rolled Steel
InsulationVacuum Insulation Panels (VIP) + Water-Blown Polyurethane Foam (SNAP-compliant)
InteriorOptional Stainless Steel
Exterior FinishPainted Cold Rolled Steal
Shelves3 Adjustable, 1 Fixed
Certifications/CompliancecULus, CE, ENERGYSTAR, EPA SNAP
StyleUpright
DisplayTouch-Screen LCD
Casters4 All-Direction Casters with Locks
Ports2 Rear Access Ports
Product LineTSX Universal Series
WarrantyWarranty varies by country, please contact us for details
ACT LabelYes
DefrostManual
ModelGeneral Purpose Ultra-Low Freezer 600
Peak Variation at -80C3.6
Water CooledNot Available
Dimensions (DxWxH) Exterior37.6 x 39.6 x 78 in. (95.5 x 100.6 x 198.1 cm)
Dimensions (D x W x H) Interior28.3 x 34.4 x 51.2 in. (71.9 x 87.3 x 130.1 cm)
Depth (English) Exterior37.6 in.
Depth (English) Interior28.3 in.
Depth (Metric) Exterior95.5 cm
Depth (Metric) Interior71.9 cm
Monitoring OptionsAudible and Visual High and Low Temperature Alarms, Wireless
Access SecurityStandard Key Lock, Padlock Loop, Setpoint and Settings Security (Optional NFC Key Card Access)
On-Board DataloggingTemperature, Events, Diagnostics
Setpoint Security2 Modes with Full Password or PIN options
Backup SystemsOptional CO2 or LN2
ConnectivityThermo Scientific™ Instrument Connect Compatible
Smart View CompatibilityOnboard accessory 5 V and POE outlets
Amperage8.9-4.9 A
Plug TypeNEMA 5-15
Voltage100/230 V
Electrical Requirements100/230 V, 50/60 Hz
Frequency50/60 Hz
PhaseSingle
WattageCompressor 938 W
Energy Usage0.27 kWh/day/cu.ft. (ENERGY STAR)
Green FeaturesACT Ecolabel (Gold), ENERGY STAR
Voltage Compensation±10%
Vial Capacity60,000 x 2 mL
CompressorV-drive
RefrigerantR290 (1st stage), R170 + R290 mix (2nd stage)
Holds2 in. Boxes (600)
Rack Capacity24 Racks
Unit SizeEach