ZERO Prefilled Formalin Container, 60 mL
ZERO Prefilled Formalin Container, 60 mL
Actual product may vary
ZERO Prefilled Formalin Container, 60 mL
Meccanica GM Divisione Medicale ZERO Prefilled Formalin Container, 60 mL protects against exposure to harmful chemicals and vapors during routine fixation of specimens in formalin.
Have Questions?
Catalog NumberColor
TSS6015Pink
Catalog number TSS6015
Price (SGD)
539.00
EA
Add to cart
Color:
Pink
Price (SGD)
539.00
EA
Add to cart
  • Reduces or eliminates exposure to formalin and harmful vapors
  • Comes in a variety of colored caps for easy specimen identification
  • Security seal
  • Dual chamber keeps formalin trapped until container is activated
Specifications
Product TypePre-filled Specimen Container
Closure IncludedYes
GraduatedNo
MaterialPolypropylene
ShapeCylindrical
SterilityNon-sterile
Capacity (Metric)60 mL
Closure MaterialPlastic
Closure TypeThreaded Cap
ColorPink
Diameter (Metric)4.445 cm
Unit SizeEA