Type I Fiberglass Step Ladder
Type I Fiberglass Step Ladder
Actual product may vary
Type I Fiberglass Step Ladder
Features a HolsterTop™, pinch-proof spreaders, traction-tred slip-resistant steps, and EDGE bracing for increased protection against damage.
Have Questions?
Catalog NumberLength (English)
TSS600560 in.
Catalog number TSS6005
Price (SGD)
539.00
EA
Add to cart
Length (English):
60 in.
Price (SGD)
539.00
EA
Add to cart
HolsterTop™ with Lock-In System secures tools to increase productivity.
  • Sturdy paint can hanger on back side of top
  • Braces on top step, second step and top horizontal
  • External Pinch-Proof™ spreaders
  • Slip-resistant Traction-Tred™ steps
  • The Lock-In System lets you customize your ladder top and safely secure your tools
  • EDGE™ molded brace and foot pad combination provides enhanced bracing strength and increased protection against damage
  • Slip-resistant foot pads attached to the EDGE™ structure
Specifications
Certifications/ComplianceANSI-A14-2-2007, OSHA
Dimensions (L x W x H)60 x 20.5 x 6.25 in.
Height (English)6 ft.
Length (English)60 in.
Model6000 Series
Quantity1 Ea.
Width (English)20.5 in.
Load Capacity (English)250 lb.
Load Capacity (Metric)113.4 kg
MaterialFiberglass
Product TypeFolding Step Ladder
Unit SizeEA