4D Series Dry Microsphere Size Standards
Actual product may vary
Thermo Scientific
4D Series Dry Microsphere Size Standards
Calibrate airborne optical particle counters to comply with ISO 21501-4 with Thermo Scientific™ 4D Series Dry Microsphere Size Standards.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)
TSS4D-1010 μm
TSS4D-055 μm
Catalog number TSS4D-10
Price (SGD)
1,523.00
Each
Diameter (Metric):
10 μm
Price (SGD)
1,523.00
Each
Calibrate airborne optical particle counters (OPC) to comply with ISO 21501-4 regulations with the Thermo Scientific™ 4D Series Dry Microsphere Size Standards. Ideal for instrument manufacturers, as well as QA/QC managers in pharmaceutical, biotech and electronics manufacturing, these microsphere reference beads feature a calibration uncertainty—the range of values within which the actual mean diameter will lie—that is less than or equal to 2.5% of the mean diameter.
The diameter is traceable to the National Institute of Standards and Technology (NIST), and the product’s Certificate of Calibration and Traceability provides the documentation needed for QC/regulatory programs and audits.
  • Mean size standard uncertainty of ≤2.5% enables calibration of air particle counters to ISO
  • Superior uniformity provides exact calibration point when setting the calibration curve
  • Diameter is traceable to NIST, a known government standard
  • Certificate of Traceability provides objective evidence for QC compliance
  • Available in 3μm, 5μm, 7μm and 10μm diameters
  • Conveniently packaged in dropper-tipped bottles for direct dispensing into sampling chamber
Specifications
AdditiveTrace flow agent
Certifications/ComplianceCertificate of Calibration and Traceability to NIST, MSDS
Concentration100% solids
Contents1g
ColorWhite
For Use With (Application)Airborne Optical Particle Counter Calibration
MaterialPolystyrene Divinylbenzene (PS-DVB)
Product TypeSize Standard Particles
Refractive Index1.59 at 589nm (25°C)
Diameter (Metric)10 μm
Unit SizeEach
Contents & Storage
Unless otherwise stated, store at ambient room temperature. Store upright; keep bottle tightly sealed. Mix product with gentle inversion by hand or vortex mixer.