4000 Series Monosized Particles
4000 Series Monosized Particles
Actual product may vary
Thermo Scientific
4000 Series Monosized Particles
Minimize response of analytic systems to shape effects with monosized particles.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)
TSS4011A1.1 μm
TSS4009A1.0 μm
FIS09-980-0401.0 μm
TSS4010A1.0 μm
TSS4016A1.6 μm
TSS4025A2.5 μm
TSS4202A2 μm
TSS4203A3 μm
FIS09-980-0495 μm
TSS4205A5 μm
TSS4210A10 μm
4215A15 μm
TSS4215A15 μm
TSS4220A20 μm
FIS09-980-05825 μm
4250A50 μm
FIS09-980-06150 μm
TSS4250A50 μm
TSS4280A80 μm
4310A100 μm
4314A140 μm
4316A160 μm
TSS4320A200 μm
4330A300 μm
4350A500 μm
4375A750 μm
Catalog number TSS4011A
Price (SGD)
1,154.00
EA
Add to cart
Diameter (Metric):
1.1 μm
Price (SGD)
1,154.00
EA
Add to cart
Minimize response of analytic systems to shape effects with Thermo Scientific™ 4000 Series Monosized Particles, excellent for use with any application that requires a NIST™ traceable size standard with a very narrow standard deviation.
Features:
  • Particle spheres are used instead of irregularly shaped particles.
  • Mean diameters have been calibrated with Thermo Fisher Scientific™'s NIST™ traceable microscopy methods.
  • Size distribution and uniformity are measured by electrical resistance analysis or optical microscopy.
  • 4000 Series particles from 1 to 160μm are made from polystyrene (PS)
  • 4000 Series particles from 200 to 1000μm are made from polystyrene crosslinked with divinylbenzene (PSDVB).
Ordering Information:
Products from 1 to 160μm are packaged as aqueous suspensions in 15mL dropper-tipped bottles, at an optimum concentration for dispersion, handling and dilution. Diameters of 200μm and larger are packaged as dry spheres. Packages are lot-numbered for convenient technical service and support after the sale.
Thermo Fisher Scientific™ has its own engineering, production and metrology facilities for better quality control and response to customer needs. In addition to this line of polymer size standards, we also offer calibration standards in silica and glass materials. Ask our technical service department for information about specific applications or other product needs not listed here.
Includes:
Certificate of Calibration and Traceability to NIST, which includes a description of the calibration method and its uncertainty; Table of chemical and physical properties; Material Safety Data Sheet
Specifications
AdditiveContains trace amount of surfactant to inhibit agglomeration and promote stability
Certifications/ComplianceEach package of standards contains a Certificate of Calibration and Traceability to NIST which includes a description of the calibration method and its uncertainty, and a table of chemical and physical properties.
Concentration1.00% solids
ContentsPolymer particles in water
DescriptionNIST traceable particles that provide calibration of laser particle counters
ColorWhite or Colorless
For Use With (Application)Narrow Size Standard Deviation Applications
MaterialPolystyrene
Product TypeSize Standard Particles
IncludesCertificate of Calibration and Traceability to NIST
Particle Density1.05g/cm³
Quantity15 mL
Refractive Index1.59 at 589nm (25°C)
Size Distributions = 0.012µm, 1.1% CV
StabilityProduct stable for a minimum of 24 months
Diameter (Metric)1.1 μm
Unit SizeEA
Contents & Storage
Store in a tightly closed container, between 4° to 30°C. Keep bottle upright and tightly sealed. Mix product with gentle inversion by hand or vortex mixer.