Search Thermo Fisher Scientific
Catalog Number | Model | Description | Capacity (Metric) |
---|---|---|---|
TSGP05 | Precision GP 05 | 5L General Purpose Water Bath | 5 L |
FKS15395837 | Precision GP 02 | 2L General Purpose Water Bath | 2 L |
TSGP02 | Precision GP 02 | 2L General Purpose Water Bath | 2 L |
TSPTSGP02 | Precision GP 02 | 2L General Purpose Water Bath | 2 L |
TSPTSGP05 | Precision GP 05 | 5L General Purpose Water Bath | 5 L |
TSGP10 | Precision GP 10 | 10L General Purpose Water Bath | 10 L |
TSPTSGP10 | Precision GP 10 | 10L General Purpose Water Bath | 10 L |
TSGP20 | Precision GP 20 | 20L General Purpose Water Bath | 20 L |
TSPTSGP20 | Precision GP 20 | 20L General Purpose Water Bath | 20 L |
TSGP28 | Precision GP 28 | 28L General Purpose Water Bath | 28 L |
TSPTSGP28 | Precision GP 28 | 28L General Purpose Water Bath | 28 L |
TSGP15D | Precision GP 15D | 5L & 10L Dual General Purpose Water Bath | 15 L |
TSPTSGP15D | Precision GP 15D | 5L & 10L Dual General Purpose Water Bath | 15 L |