QSP Low Retention Non-Filtered Pipette Tips
Actual product may vary
Thermo Scientific
QSP Low Retention Non-Filtered Pipette Tips
Low retention pipette tips with a hydrophobic inner surface to reduce liquid retention and maintain pipetting precision.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Tip StyleFormatSterility
TLR112NXRLS-Q1000 μLExtended LengthRefillSterile
TLR102-Q10 μLExtended LengthBulkNon-sterile
TLR102R-Q10 μLExtended Length, ConicalHinged RackNon-sterile
TLR102RS-Q10 μLExtended LengthHinged RackSterile
TLR102RL-Q10 μLExtended LengthRefillNon-sterile
TLR102RLS-Q10 μLExtended LengthRefillSterile
TLR070-Q200 μLBeveledBulkNon-sterile
TLR070R-Q200 μLBeveledHinged RackNon-sterile
TLR070RS-Q200 μLBeveledHinged RackSterile
TLR070RL-Q200 μLBeveledRefillNon-sterile
TLR070RLS-Q200 μLBeveledRefillSterile
TLR106-Q300 μLMicroPointBulkNon-sterile
TLR106RS-Q300 μLMicro PointHinged RackSterile
TLR112NXL-Q1000 μLExtended LengthBulkNon-sterile
TLR112NXLR-Q1000 μLExtended LengthHinged RackNon-sterile
TLR112NXLRS-Q1000 μLExtended LengthHinged RackSterile
TLR112NXLRL-Q1000 μLExtended LengthRefillNon-sterile
Catalog number TLR112NXRLS-Q
HSN 39269099
Price (SGD)
77.00
PK
Volume (Metric):
1000 μL
Tip Style:
Extended Length
Format:
Refill
Sterility:
Sterile
Price (SGD)
77.00
PK
Dispensing certain liquids can form a film on your tip, which can affect assay accuracy and precision. Thermo Scientific™ QSP low retention pipette tips offer a solution with a hydrophobic inner surface that reduces liquid retention.
Featuring:
  • Minimize the formation of films on inside of tip
  • Reduce sample retention
  • Improve accuracy and precision
  • Improve sample recovery
  • High clarity pipette tips
  • Variety of packaging options, including environmentally friendly tip reload systems
Clean Claims:
QSP Pipette Tips are free of:
  • Non-Sterile:
    • RNase/DNase
    • DNA
    • Pyrogen
  • E-Beam Sterilized:
    • RNase/DNase
    • DNA
    • Pyrogen/Endotoxin
    • Bioburden
    • PCR inhibitors
    • Endotoxin ATP
To order a clean claim lot-specific certification, contact your local dealer with the Cat. No and Lot No.
Recommend using with Finnpipette Digital pipette, but also compatible with other brands of pipettes.
Specifications
DescriptionQSP 1000, Non-Filtered
Volume (Metric)1000 μL
FilteredNon-filtered
SterilitySterile
ColorNatural
ModelQSP 1000
Length (Metric)8.9 cm
Clean ClaimsDNase, RNase, DNA, ATP, PCR Inhibitor, and Pyrogen Free
FormatRefill
Tip StyleExtended Length
For Use With (Equipment)Universal Fit
Product LineQSP
Unit SizePK