QSP Low Retention Non-Filtered Pipette Tips
Actual product may vary
QSP Low Retention Non-Filtered Pipette Tips
Low retention pipette tips with a hydrophobic inner surface to reduce liquid retention and maintain pipetting precision.
Have Questions?
Catalog NumberVolume (Metric)Tip StyleFormatSterility
TLR112NXLRL-Q1000 μLExtended LengthRefillNon-sterile
Catalog number TLR112NXLRL-Q
HSN 39269099
Price (SGD)
76.00
PK
Volume (Metric):
1000 μL
Tip Style:
Extended Length
Format:
Refill
Sterility:
Non-sterile
Price (SGD)
76.00
PK
Dispensing certain liquids can form a film on your tip, which can affect assay accuracy and precision. Thermo Scientific™ QSP low retention pipette tips offer a solution with a hydrophobic inner surface that reduces liquid retention.
Featuring:
  • Minimize the formation of films on inside of tip
  • Reduce sample retention
  • Improve accuracy and precision
  • Improve sample recovery
  • High clarity pipette tips
  • Variety of packaging options, including environmentally friendly tip reload systems
Clean Claims:
QSP Pipette Tips are free of:
  • Non-Sterile:
    • RNase/DNase
    • DNA
    • Pyrogen
  • E-Beam Sterilized:
    • RNase/DNase
    • DNA
    • Pyrogen/Endotoxin
    • Bioburden
    • PCR inhibitors
    • Endotoxin ATP
To order a clean claim lot-specific certification, contact your local dealer with the Cat. No and Lot No.
Recommend using with Finnpipette Digital pipette, but also compatible with other brands of pipettes.
Specifications
DescriptionQSP 1000, Non-Filtered
Volume (Metric)1000 μL
FilteredNon-filtered
SterilityNon-sterile
ColorNatural
ModelQSP 1000
Length (Metric)8.9 cm
Clean ClaimsDNase, RNase, DNA, and Pyrogen Free
FormatRefill
Tip StyleExtended Length
For Use With (Equipment)Universal Fit
Product LineQSP
Unit SizePK