Beveled Pipette Tips, 2-20 μL, Filtered, Clear, Sterile
Actual product may vary
Beveled Pipette Tips, 2-20 μL, Filtered, Clear, Sterile
Filter and non-filter pipette tips in a complete portfolio for your evolving liquid transfer needs.
Have Questions?
Catalog number TF113-20-Q
HSN 84799090
Price (SGD)
41.40
Special offer
Ends: 30-Nov-2026
138.00
Save 96.60 (70%)
PK
Price (SGD)
41.40
Special offer
Ends: 30-Nov-2026
138.00
Save 96.60 (70%)
PK
Thermo Scientific™ QSP filter and non-filtered pipette tips are made from 100% virgin polypropylene utilizing state of the art molds and molding methods. Pipette tips are molded with high clarity resin and many include graduation marks and micro point design. Packaging options range from bulk to rack and rack sterile and include environmentally friendly options like tip reload systems and stack racks.
Featuring:
  • Micro point design tip orifice
  • High clarity resin
  • Graduation marks
  • Universal fit
  • Small micron filter
  • Modern and robust tip rack design
Clean Claims:
QSP Pipette Tips are free of:
  • Non-Sterile:
    • RNase/DNase
    • DNA
    • Pyrogen
  • E-Beam Sterilized:
    • RNase/DNase
    • DNA
    • Pyrogen/Endotoxin
    • Bioburden
    • PCR inhibitors
    • Endotoxin ATP
To order a clean claim lot-specific certification, contact your local dealer with the Cat. No and Lot No.
Recommend using with Finnpipette Digital pipette, but also compatible with other brands of pipettes.
Specifications
DescriptionQSP 20, Filtered
Volume (Metric)20 μL
FilteredFiltered
SterilitySterile
ColorNatural
Length (Metric)5.1 cm
Clean ClaimsDNase, RNase, DNA, ATP, PCR Inhibitor, and Pyrogen Free
FormatHinged Rack
Tip StyleBeveled
Quantity4800/Cs.
Green FeaturesACT Ecolabel (Silver)
For Use With (Equipment)Universal Fit
GraduatedYes
Product LineQSP
MaterialVirgin Polypropylene
Unit SizePK