Stainless Steel Upright Freezer Rack for Cryogenic Boxes
Actual product may vary
Tenak
Stainless Steel Upright Freezer Rack for Cryogenic Boxes
Use Tenak™ Stainless Steel Upright Freezer Racks for Cryogenic Boxes for your cryogenic storage needs. A variety of stainless steel racks accommodate box heights and quantities for laboratory freezers.
Have Questions?
Catalog NumberHoldsArray
TENTE23154Boxes (25)5 x 5
Catalog number TENTE23154
Price (SGD)
-
Holds:
Boxes (25)
Array:
5 x 5
  • Suitable for all brands of upright freezers
  • Stainless steel
  • For cryogenic boxes of various capacities
Specifications
Cell DividersYes
HoldsBoxes (25)
MaterialStainless Steel
Positions25
For Use With (Application)Fit into any freezer brand and model
For Use With (Equipment)Upright Freezers
No. per Pack1
Quantity1 Ea.
Depth (Metric)688 mm
Product TypeStandard Rack
Shelves25
Height (Metric)276 mm
Array5 x 5
Width (Metric)139 mm
Unit SizeEach