▸
Boxes
▸
Cryogenic and Freezer Boxes
▸
Polypropylene Cryobox, 50 mm Height
Actual product may vary
Tenak
Polypropylene Cryobox, 50 mm Height
Cryobox with cell codes printed on the lid for easy identification.
Have Questions?
Catalog Number
Color
TENTE22315
Red
Catalog number
TENTE22315
Price (SGD)
-
Color:
Red
Constructed of polypropylene
Can be used in temperatures as low as -90°C
Autoclavable at 121°C for 20 minutes
Boxes have a fixed grid divider with 9 x 9 cells and are suited for 1.2 mL and 2 mL cryogenic vials
Specifications
Color
Red
Material
Polypropylene
Withstand Temperature (Metric)
As low as -90°C
Dimensions (L x W x H)
130 x 130 x 50 mm