Cardboard Cryoboxes
Actual product may vary
Tenak
Cardboard Cryoboxes
Combine Tenak™ Cardboard Cryoboxes with different grid inserts for tubes from 6 to 28.5mm in diameter. Available in many colors, the highly flexible;boxes can be used for larger laboratory containers when removing the grid.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorDimensions (L x W x H)Includes
TENTE22240White100 x 100 mmDividers
TENTE22010Blue133 x 133 x 50 mmDividers
TENTE22015Red133 x 133 x 50 mmDividers
TENTE22235White75 x 75 mmDividers
TENTE22025Yellow133 x 133 x 50 mmDividers
FKS15220524White133 x 133 x 75 mm
TENTE22050White133 x 133 x 75 mm
FKS15260534White133 x 133 x 100 mmDividers
TENTE22100White133 x 133 x 100 mmDividers
TENTE22230White133 x 133 x 52 mm9 x 9 Divider
TENTE22102Red133 x 133 x 100 mmDividers
Catalog number TENTE22240
Price (SGD)
-
Color:
White
Dimensions (L x W x H):
100 x 100 mm
Includes:
Dividers
  • Cardboard
  • Temperature resistant down to -80°C
  • Ideal for ultra-low-temperature storage
  • The required divider height depends on the cryobox height:
    • 22 mm for 32 mm and 40 mm boxes
    • 30 mm for 50 mm boxes
    • 40 mm for 75 mm boxes
    • 65 mm for 100 mm and 130 mm boxes
Specifications
ColorWhite
MaterialCardboard
ShapeSquare
TypeCryo Box
For Use With (Application)Cryogenic Storage of Liquids
IncludesDividers
Quantity36 Pack
Dimensions (L x W x H)100 x 100 mm
Width (Metric)100 mm
Height (Metric)100 mm
Weight (Metric)4.4 kg
Unit SizePack