Search Fisher Scientific
| Catalog Number | Holds | Tube Array |
|---|---|---|
| TENTE22142 | Vials with 15 mm O.D. | 8 x 8 cells |
| FKS15240554 | Vials with 31 mm O.D. | 4 x 4 cells |
| TENTE22148 | Vials with 31 mm O.D. | 4 x 4 cells |
| FKS15250554 | Vials with 24 mm O.D. | 5 x 5 cells |
| TENTE22149 | Vials with 24 mm O.D. | 5 x 5 cells |
| TENTE22144 | Vials with 13 mm O.D. | 9 x 9 cells |