▸
Boxes
▸
Cryogenic and Freezer Boxes
▸
Viking Termometerfabriken™Cardboard Cryogenic Box
Actual product may vary
Tenak
Viking Termometerfabriken™Cardboard Cryogenic Box
Have Questions?
Catalog Number
Height (Metric)
TENTE22101
100 mm
Catalog number
TENTE22101
Price (SGD)
323.00
Pack
Add to cart
Height (Metric):
100 mm
Price (SGD)
323.00
Pack
Add to cart
Specifications
Color
Blue
Material
Cardboard
Shape
Square
Type
Cryo Box
Quantity
36 Pack
Length (Metric)
133 mm
Width (Metric)
133 mm
Height (Metric)
100 mm
Weight (Metric)
3.5 kg