Cardboard Cryoboxes
Cardboard Cryoboxes
Actual product may vary
Tenak
Cardboard Cryoboxes
Combine Tenak™ Cardboard Cryoboxes with different grid inserts for tubes from 6 to 28.5mm in diameter. Available in many colors, the highly flexible;boxes can be used for larger laboratory containers when removing the grid.
Have Questions?
Catalog NumberColorDimensions (L x W x H)Includes
TENTE22050White133 x 133 x 75 mmDividers
Catalog number TENTE22050
Price (SGD)
192.00
Pack
Add to cart
Color:
White
Dimensions (L x W x H):
133 x 133 x 75 mm
Includes:
Dividers
Price (SGD)
192.00
Pack
Add to cart
  • Cardboard
  • Temperature resistant down to -80°C
  • Ideal for ultra-low temperature storage
Specifications
ColorWhite
MaterialCardboard
ShapeSquare
TypeCryo Box
For Use With (Application)Cryogenic Storage of Liquids
IncludesDividers
Quantity36 Pack
Dimensions (L x W x H)133 x 133 x 75 mm
Length (Metric)133 mm
Width (Metric)133 mm
Height (Metric)75 mm
Weight (Metric)3.2 kg
Unit SizePack