TDE Series -40°C Ultra-Low Temperature Freezers
TDE Series -40°C Ultra-Low Temperature Freezers
Actual product may vary
TDE Series -40°C Ultra-Low Temperature Freezers
TDE Series -40°C Ultra-Low Temperature Freezers
TDE Series -40°C Ultra-Low Temperature Freezers
TDE Series -40°C Ultra-Low Temperature Freezers
TDE Series -40°C Ultra-Low Temperature Freezers
TDE Series -40°C Ultra-Low Temperature Freezers
TDE Series -40°C Ultra-Low Temperature Freezers
TDE Series -40°C Ultra-Low Temperature Freezers
Thermo Scientific™ TDE Series -40°C ultra-low temperature freezers feature reduced energy usage, a simplified touchscreen interface, three capacities, and an ergonomic cabinet design.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberElectrical RequirementsHoldsCertifications/Compliance
TDE60040FV230V, 50Hz2 in. Boxes (600)CE
TDE40040FV230V, 50Hz2 in. Boxes (400)CE
Catalog number TDE60040FV
Price (SGD)
23,007.00
EA
Add to cart
Electrical Requirements:
230V, 50Hz
Holds:
2 in. Boxes (600)
Certifications/Compliance:
CE
Price (SGD)
23,007.00
EA
Add to cart
Thermo Scientific™ TDE Series -40°C ultra-low temperature freezers, powered by H-drive, feature three upright models, maximizing storage capacity from 40,000 up to 60,000 2mL vials. The TDE Series is a sustainable and reliable solution with ultimate sample security and operational savings for every lab.
Performance
  • Powered by H-drive's hydrocarbon technology, these systems save up to almost 50% in energy usage and heat rejection over conventional refrigerant ultra-low freezers1
  • A quiet sound output below 53 dBA, enables the freezer to come out of the hallway and into the lab
  • Meets ±5°C validation protocols at -40°C with strong peak variation and steady-state temperature variation
  • Natural hydrocarbon refrigerants reduce system operating stresses promoting outstanding reliability and worry-free sample protection
User Friendly
  • The H-drive Information Center (HIC) combines easy to read temperature with simple capacitive touch buttons in a 5.6 inch display
  • Three capacities (400, 500 and 600 2-inch box) to accommodate a variety of storage needs and lab footprint specifications
  • All models contain four inner doors allowing for easy sample access without exposing greater freezer content to ambient conditions
  • Improved customer convenience with new ergonomically designed handle, high contrast eye-level display, and comfortable easy-access shelf locations.
  • Compatible with most racking systems including a full range of Thermo Scientific™ racks for boxes, microplates, Thermo Scientific™ Matrix™ and Nunc CryoBank™ tubes
  • Security features including alarm settings and padlock-compatible doors
Sustainability
  • Manufactured in an award-winning, zero-waste to landfill facility
  • Lower HVAC costs as a result of less heat emission into the environment
  • Natural refrigerants and water-blown foam insulation, making the Series compliant with the U.S. Environmental Protection Agency's Significant New Alternatives Policy (SNAP), the European Union's F-Gas and other sustainability standards
Footnotes:
  1. Internal data comparing 4.4kWh/day to Legacy Performance DxF40040 Model with 8.0kWh/day. Data on File
  2. Typical freezer test data based on internal testing in ambient temperature at 20°C; Typical standard validation protocol of ±5°C.
Specifications
DescriptionThermo Scientific TDE Series -40°C Ultra-Low Freezers, 600-Box Capacity, 230V, 50Hz
Capacity (English)28.8 cu. ft.
Capacity (Metric)816 L
Temperature Range-10°C to -40°C
Door StyleOuter (1), Inner (4)
No. of Doors5
InsulationVacuum Insulation Panels (VIP) + Water-Blown Polyurethane Foam (SNAP-compliant)
InteriorStainless Steel
Shelves3
Capacity (English) Shelf245 lb.
Capacity (Metric) Shelf111 kg
Certifications/ComplianceCE
StyleUpright
DisplayHIC 5.6 in. 7-segment display with capacitive touch buttons
Product LineThermo Scientific
WarrantyWarranty varies by country, please contact us for details
DefrostManual
Dimensions (DxWxH) Exterior38.5 x 39.6 x 78 in. (97.7 x 100.6 x 198.1cm)
Dimensions (D x W x H) Interior28.3 x 34.4 x 51.2 in. (71.9 x 87.3 x 130.1cm)
Monitoring OptionsAlarm, Recorder
Setpoint SecurityYes
ConnectivityRemote Alarm Connections, 4-20mA, RS485, and Dry Contacts
Smart View CompatibilitySmart-Vue Compatible
Amperage3.4/10
Plug TypeEU
Voltage230 V
Electrical Requirements230V, 50Hz
Frequency50 Hz
Vial Capacity60,000 x 2mL
CompressorH-drive
RefrigerantR290
Shipping Weight (English)765 lb.
Shipping Weight (Metric)348kg
Weight (English)654 lb.
Weight (Metric)297 kg
Holds2 in. Boxes (600)
Rack Capacity12
Unit SizeEA
Frequently asked questions (FAQs)
The low battery alarm is displaying on my ULT (Ultra-Low Temperature) freezer. Can I buy a new battery?
You can order a new battery (Cat. No. 400159) online.