Heat-Shrink Bands for Nalgene™ PETG Media Bottles
Actual product may vary
Thermo Scientific
Heat-Shrink Bands for Nalgene™ PETG Media Bottles
Ensure the integrity of bottle contents with these tamper-resistant seals. Heat-shrink bands are gamma stable and include a perforated tear strip feature for easy removal.
Have Questions?
Catalog NumberNeck Finish
TBP312160-038438-430
Catalog number TBP312160-0384
Price (SGD)
184.00
Case
Neck Finish:
38-430
Price (SGD)
184.00
Case
Ensure the integrity of bottle contents with Thermo Scientific™ Heat-Shrink Bands for Thermo Scientific™ Nalgene™ PETG Media Bottles. Simply apply the recommended application torque for Thermo Scientific Nalgene closures and slide the heat-shrink band over the closure and bottle neck. Heat the band to shrink and secure contents with a tamper-resistant seal.
  • Gamma stable bands include a perforated ”tear strip” feature for easy removal.
  • Bands available are in four sizes to mate with Nalgene PETG Square Media Bottles.
  • Bands are packed 1,000 per case (two zipper bags of 500 each).
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Warranty: 90 days
Specifications
MaterialPETG
No. per Case1000
No. per Pack1000
Torque27-33 in.-lb
Torque Wrench Fittings2195-1438
Neck Finish38-430
For Use With (Equipment)125 to 1000 mL Square Media Bottle
Product TypeHeat-shrink Band
Unit SizeCase