▸
Solvents
▸
Other Solvents
▸
Isooctane
▸
2,2,4-Trimethylpentane, Certified AR for Analysis, Fisher Chemical™
Element Search
Structure Search
Fisher Chemicals
2,2,4-Trimethylpentane, Certified AR for Analysis, Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
T360117
also known as
T/3601/17
2.5 L
Amber Glass Bottle
Catalog number
T360117
also known as
T/3601/17
HSN
29011000
Price (SGD)
-
Quantity:
2.5 L
Packaging:
Amber Glass Bottle
Request bulk or custom format
Specifications
Melting Point
-107°C
CAS
540-84-1
Color
Colorless
Quantity
2.5 L
Packaging
Amber Glass Bottle
View more