SuperNuova+™ Stirring Hotplates Series
SuperNuova+™ Stirring Hotplates Series
Actual product may vary
SuperNuova+™ Stirring Hotplates Series
SuperNuova+™ Stirring Hotplates Series
SuperNuova+™ Stirring Hotplates Series
SuperNuova+™ Stirring Hotplates Series
SuperNuova+™ Stirring Hotplates Series
Get reliable performance, dependable safety and simple operation.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTop Plate MaterialSizeElectrical RequirementsPlug Type
SP88857198Aluminum7.25 x 7.25 in.230 V 50/60 HzEU/UK/AUS/CN
SP88850195Ceramic10.25 x 10.25 in.230 V 50/60 HzEU/UK/AUS/CN
SP88857195Ceramic7.25 x 7.25 in.230 V 50/60 HzEU/UK/AUS/CN
Catalog number SP88857198
Price (SGD)
2,414.00
EA
Add to cart
Top Plate Material:
Aluminum
Size:
7.25 x 7.25 in.
Electrical Requirements:
230 V 50/60 Hz
Plug Type:
EU/UK/AUS/CN
Price (SGD)
2,414.00
EA
Add to cart
Get reliable performance, dependable safety and simple operation with our new Thermo Scientific™ SuperNuova+™ stirring hotplate series . Units are available in two sizes, including a multi-position model with optimized controls and settings for applications that demand advanced precision.
Intuitive design
  • Top plate choices in ceramic or aluminum
  • Easy-to-read LED display for heating and stirring
  • Raised display design protects electronics from spills
  • Ability to save temperature setting
  • Optional PT1000 temperature probe and splash guard protection shield
  • Includes: PT100 Temperature Probe, Rod and Clamps
Advanced technology
  • Stir Trac features innovative engineering for slow-speed stirring, consistent speed control, and stronger magnetic couplng
  • Hot Top warning system protects from accidental burns with prominent display when heating surface is above 50°C (122°F)
Warranty: 3 year warranty
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Top Plate MaterialAluminum
Voltage230 V
IncludesPower Cord
Shipping Weight (English)12.1 lb.
Shipping Weight (Metric)5.5 kg
Electrical Requirements230 V 50/60 Hz
Length (English)13 in.
Size7.25 x 7.25 in.
Height (Metric)9.7 cm
Temperature (Metric)Ambient to 300°C
Height (English)3.8 in.
Frequency50/60 Hz
Width (Metric)20.8 cm
Length (Metric)33 cm
Width (English)8.2 in.
Weight (English)12.1 lb.
Weight (Metric)5.5 kg
TypeStirring Hotplate, Stirrer
ShapeSquare
Plug TypeEU/UK/AUS/CN
Stirring Range50 to 1500 rpm
Warranty3 Years
Dimensions (LxWxH)7.25 in. x 7.25 in.
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
DisplayLED
Unit SizeEA