Isotemp™ Hotplate Stirrer, Ambient to 540°C, Ceramic
Isotemp™ Hotplate Stirrer, Ambient to 540°C, Ceramic
Actual product may vary
Isotemp™ Hotplate Stirrer, Ambient to 540°C, Ceramic
Stirring Range: 50 to 1500 rpm, Shape: Square, Size: 4.25 x 4.25 in., Plug Type: EU and UK
Have Questions?
Catalog NumberPlug Type
SP88854205UK/EU/CN
Catalog number SP88854205
Price (SGD)
978.00
EA
Add to cart
Plug Type:
UK/EU/CN
Price (SGD)
978.00
EA
Add to cart
Experience precise stirring control, exceptional safety and temperature performance for your routine protocols.
Advanced technology:
  • Slow-speed stirring, consistent speed control, and strong magnetic coupling
  • Temperature adjustable in 1 degree increments
  • HOT TOP warning system designed to protect from accidental burns with prominent display when heating surface is above 50°C (122°F)
  • Automatic over-temperature protection
Flexible, robust design:
  • Top plate choices in ceramic or aluminum
  • Raised display design protects electronics from spills
  • Optional temperature probe and splash guard protection shield
  • Variable sizes for different sample volumes
  • Includes stir bar
  • EU and UK plug
Specifications
Top Plate MaterialCeramic
Voltage230 V
DescriptionHotplate Stirrer
IncludesStir Bar
Electrical Requirements230 V 50/60 Hz
Length (English)5 in.
Size4.25 x 4.25 in.
Height (Metric)9.1 cm
Temperature (Metric)Ambient to 540°C
Height (English)3.6 in.
Frequency50/60 Hz
Width (Metric)25.4 cm
Length (Metric)12.7 cm
Width (English)10 in.
TypeStirring Hotplate
ShapeSquare
Plug TypeUK/EU/CN
Max. Container Size1 L
Stirring Range50 to 1500 rpm
Warranty3 Years
Dimensions (LxWxH)10 x 5 x 3.6 in. (25.4 x 12.7 x 9.1 cm)
Certifications/ComplianceCE, RoHS Compliant
Unit SizeEA