Corn Oil, NF, Spectrum™ Chemical
Spectrum Chemical Mfg. Corp.
Corn Oil, NF, Spectrum™ Chemical
CO136, 8001-30-7
Have Questions?
Catalog NumberQuantityPackaging
SCMCO136-500MLGL500 mLDrum
Catalog number SCMCO136-500MLGL
Price (SGD)
-
Quantity:
500 mL
Packaging:
Drum
Request bulk or custom format
Specifications
Chemical Name or MaterialCorn Oil
CAS8001-30-7
CAS Max %100%
PackagingDrum
Quantity500 mL
View more
Spectrum™ Chemical Corn Oil, NF its uses include as a solvent vehicle for topical preparations and as a glidant excipient for oral dosage tablets and capsules. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.