▸
Stoppers and Closures
▸
Bottle Caps
▸
DURAN™ GL 45 Red Pouring Rings For Screw Caps
Actual product may vary
Schott Duran
DURAN™ GL 45 Red Pouring Rings For Screw Caps
ETFE pouring ring
Have Questions?
Catalog Number
Thread Size (Metric)
SCH292442802
9 mm
Catalog number
SCH292442802
Price (SGD)
47.00
Case
Add to cart
Thread Size (Metric):
9 mm
Price (SGD)
47.00
Case
Add to cart
Chemical resistant
Autoclavable
Max. temp.: 180°C
Specifications
Color
Red
Height (Metric) With Closure
4mm
Material
ETFE
Thread Size (Metric)
9 mm
Neck Finish
9 mm
Product Type
Pouring Ring
Autoclavable
Autoclavable