DURAN™ Weighing Bottle, with ground lid, low form
DURAN™ Weighing Bottle, with ground lid, low form
Actual product may vary
DURAN™ Weighing Bottle, with ground lid, low form
Weighing glasses are suitable for weighing substances. The matching lid prevents unintentional distribution of the weighing product.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
SCH24210320230 mL
FKS1174094415 mL
FKS1170096430 mL
Catalog number SCH242103202
Price (SGD)
824.00
CS
Add to cart
Capacity (Metric):
30 mL
Price (SGD)
824.00
CS
Add to cart
In order to carry out reactions in a controlled manner, it is essential to know the quantities of substances required and to weigh the corresponding substances accurately. A weighing glass can be used for weighing. The associated lid allows the weighing pan to be closed and thus prevents, for example, turbulence of the weighed-in solids during transport. Since even the smallest contamination of the weighing glass can have an effect on the weighing inheritance, the glass in the area of the ground joint and the lid knob are so pronounced that they can be grasped and moved with forceps or tweezers.
  • Close-fitting lid prevents any sample loss during transport after weighing
  • Fired-on, highly durable white ceramic
Specifications
Capacity (Metric)30 mL
DescriptionLow form
For Use With (Application)Weighing; Storage and transport; Available in low and high forms
MaterialBorosilicate Glass (3.3)
Quantity10/Pk.
AutoclavableAutoclavable
Diameter (Metric)54 mm
Unit SizeCS