DURAN™ GL 45 Laboratory Glass Bottles With PP Screw Cap and Pouring Ring
Actual product may vary
Duran
DURAN™ GL 45 Laboratory Glass Bottles With PP Screw Cap and Pouring Ring
With proven DURAN™ properties. Available as bottle only and completed bottle with blue screw cap (PP1, integral lip seal) and pouring ring (PP1) for drip-free pouring and clean, safe working.
Have Questions?
Catalog NumberCapacity (Metric)
SCH21801865810000 mL
Catalog number SCH218018658
Price (SGD)
537.00
Case
Capacity (Metric):
10000 mL
Price (SGD)
537.00
Case
  • Service temperature level of screw cap and pouring ring: +140 °C
  • Typical applications: storage, sample preparation, transport
  • Select sizes available with retrace code
Specifications
Capacity (Metric)10000 mL
ColorClear
MaterialGlass
Thread Size (Metric)GL 45
Height (Metric)415 mm
Product TypeLaboratory Bottle
Diameter (Metric)227 mm
Unit SizeCase