DURAN™ Low Form Beakers with Spout
Actual product may vary
DURAN™ Low Form Beakers with Spout
DURAN™ Low Form Beakers with Spout
Schott Duran
DURAN™ Low Form Beakers with Spout
Uniform wall thickness distribution makes these beakers ideal for heating applications.
Have Questions?
Catalog NumberCapacity (Metric)
SCH2110663012000 mL
Catalog number SCH211066301
Price (SGD)
278.00
Case
Capacity (Metric):
2000 mL
Price (SGD)
278.00
Case
  • Easy-to-read scale and large labelling field for easy marking in fired-on, highly durable, white ceramic
  • Spout for clean pouring
Specifications
Capacity (Metric)2000 mL
MaterialBorosilicate Glass
Height (Metric)185 mm
Quantity10/Pk.
TypeBeaker
Unit SizeCase