SuperNuova+™ Stirrer Series
SuperNuova+™ Stirrer Series
Actual product may vary
SuperNuova+™ Stirrer Series
SuperNuova+™ Stirrer Series
SuperNuova+™ Stirrer Series
Get reliable performance, dependable safety and simple operation with this stirrer series.
Have Questions?
Catalog NumberHeating Surface Area (English)Top Plate MaterialElectrical RequirementsPlug Type
S888571957.25 x 7.25 in.Ceramic230V 50/60HzAUS/CN/EU/UK
Catalog number S88857195
Price (SGD)
-
Heating Surface Area (English):
7.25 x 7.25 in.
Top Plate Material:
Ceramic
Electrical Requirements:
230V 50/60Hz
Plug Type:
AUS/CN/EU/UK
Get reliable performance, dependable safety and simple operation with the Thermo Scientific™ SuperNuova+™ Stirrer series. Units are available in two sizes, including a multi-position model with optimized controls and settings for applications that demand advanced precision.
Intuitive design
  • Top plate choices in ceramic or aluminum
  • Easy-to-read LED display for heating and stirring
  • Raised display design protects electronics from spills
  • Ability to set programs (ramp/dwell) as well as save settings
  • Optional PT1000 temperature probe and splash guard protection shield
Advanced technology
  • Stir Trac features innovative engineering for slow-speed stirring, consistent speed control, and stronger magnetic couplng
  • Hot Top warning system protects from accidental burns with prominent display when heating surface is above 50°C (122°F)
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement. Warranty: Subject to Thermo Fisher Scientific standard limited warranty. See thermoscientific.com or your sales representative for details.
Specifications
Speed50 to 1500 rpm
Top Plate MaterialCeramic
Capacity4 L
DisplayLED
ShapeSquare
Voltage230 V
Dimensions (L x W x H)13 x 8.2 x 3.8 in. (33 x 20.8 x 9.7 cm)
Heating Surface Area (English)7.25 x 7.25 in.
Heating Surface Area (Metric)18.4 x 18.4 cm
Electrical Requirements230V 50/60Hz
Frequency50/60 Hz
Plug TypeAUS/CN/EU/UK
Certifications/ComplianceCCSAus, CE
TypeStirrer
Shipping Weight (English)12.1 lb.
Shipping Weight (Metric)5.5 kg
Unit SizeEA