Super-Nuova™ Multi-Position Digital Stirrers
Super-Nuova™ Multi-Position Digital Stirrers
Actual product may vary
Super-Nuova™ Multi-Position Digital Stirrers
Carry out solubility studies, digestion, titration and enzymatic studies with Thermo Scientific™ Super-Nuova™ Multi-Position Digital Stirrers.
Have Questions?
Catalog NumberElectrical Requirements
S136030-33Q220/240V 50/60Hz
Catalog number S136030-33Q
Price (SGD)
4,429.00
EA
Add to cart
Electrical Requirements:
220/240V 50/60Hz
Price (SGD)
4,429.00
EA
Add to cart
Carry out solubility studies, digestion, titration and enzymatic studies with Thermo Scientific™ Super-Nuova™ Multi-Position Digital Stirrers. Offering four individually-controlled stirring positions, ceramic tops and 4-function memory, these stirrers are designed for stronger coupling and stirring control with advanced safety features. Ideal for labs with multiple users.
Key Features
  • Thermo Scientific™ Stir Trac™ stir control provides improved slow-speed stirring, consistent speed, and strong magnetic coupling — reducing the risk of stir bar decoupling
  • Microprocessor control ensures accurate and stable setpoints for stirring speed
  • Stir Trac braking feature brings stir bar to an immediate stop for quick flask removal
  • Single control knob controls speed; lock feature prevents accidental changes
  • Digital rpm display
  • Clean easily and resist alkalis and acids
  • Seamless, reflective white ceramic surface aids sample visibility
  • Stirring adjustable in units of 1rpm
  • RS-232 port outputs time count, stir speed (set point) directly to your PC
Safety Features
  • Rugged low-profile cast-aluminum body provides stability and durability
  • Unit design protects internal components from accidental spills
  • Walk-away timer shuts off stirring after 1, 2, 4, 8, or 12 hr.
Includes:
  • Detachable line cord and plug
Specifications
Speed50 to 1200 rpm
Stirring Positions5
Top Plate MaterialCeramic
DisplayDigital
ShapeSquare
Voltage220/240 V
Dimensions (L x W x H)16.2 x 11.3 x 4 in. (41.1 x 28.7 x 10.2 cm)
Height (English)4 in.
Height (Metric)10.2 cm
Length (English)16.2 in.
Length (Metric)41.1 cm
Top Plate Length (English)10.25 in.
Top Plate Width (English)10.25 in.
Width (English)11.3 in.
Width (Metric)28.7 cm
Electrical Requirements220/240V 50/60Hz
Frequency50/60 Hz
Certifications/ComplianceCCSAus, CE
Description4 Position, 220-240V
MaterialAluminum
TypeStirrer
Shipping Weight (English)20.5 lb.
Shipping Weight (Metric)9.3 kg
Unit SizeEA