Savant™ SpeedVac™ Rotors for Large Capacity Concentrators
Savant™ SpeedVac™ Rotors for Large Capacity Concentrators
Actual product may vary
Savant™ SpeedVac™ Rotors for Large Capacity Concentrators
SpeedVac Rotors for Large Capacity Concentrators for use with Savant SpeedVac Concentrators.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberModelDescriptionHolds
RH200-12
also known as AVL
RH200-12Microtubes: 200 x 1.5mL; Glass and Plastic Tubes: 200 (12 x 75mm), Large capacity models1.5 mL Microtube, 12 x 75 mm, 13 x 100 mm Glass and Plastic Tubes
MPTR8-210MPTR8-210Microwell Plates: 8 Deep-well, Large capacity models8 Microwell Plates
MPTR12-210
also known as AVL
MPTR12-210Microwell Plates: 12 Shallow-well, Large capacity models12 Deep-well Plates
Catalog number RH200-12
also known as AVL
Price (SGD)
3,167.00
EA
Add to cart
Model:
RH200-12
Description:
Microtubes: 200 x 1.5mL; Glass and Plastic Tubes: 200 (12 x 75mm), Large capacity models
Holds:
1.5 mL Microtube, 12 x 75 mm, 13 x 100 mm Glass and Plastic Tubes
Price (SGD)
3,167.00
EA
Add to cart
SpeedVac Rotors for Large Capacity Concentrators for use with Savant SpeedVac Concentrators.
Various size rotors made from either plastic or aluminum to fit various sample formats of your choice.
  • Easy to use
  • Multi-size tube holes accommodate different sample sizes
  • Require no additional accessories
  • Fit directly into your concentrator
Specifications
DescriptionMicrotubes: 200 x 1.5mL; Glass and Plastic Tubes: 200 (12 x 75mm), Large capacity models
MaterialGlass (Tube), Plastic (Tube)
TypeRotor
For Use With (Equipment)SPD210, SPD2030, SPD300DDA (Requires RAD200)
Holds1.5 mL Microtube, 12 x 75 mm, 13 x 100 mm Glass and Plastic Tubes
ModelRH200-12
Rotor SystemTraditional
Unit SizeEA