Savant™ DNA SpeedVac™ Concentrators Rotors
Savant™ DNA SpeedVac™ Concentrators Rotors
Actual product may vary
Savant™ DNA SpeedVac™ Concentrators Rotors
Savant™ DNA SpeedVac™ Concentrators Rotors
Savant™ DNA SpeedVac™ Concentrators Rotors
Savant™ DNA SpeedVac™ Concentrators Rotors
Savant™ DNA SpeedVac™ Concentrators Rotors
Configure Thermo Scientific™ Savant™ DNA SpeedVac™ Concentrators Rotors with Savant DNA SpeedVac Concentrators.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberHolds
RD7272
RD2MP
also known as AVL
2 Shallow-well Plates
RD48
also known as AVL
48
Catalog number RD72
Price (SGD)
1,763.00
EA
Add to cart
Holds:
72
Price (SGD)
1,763.00
EA
Add to cart
Configure Thermo Scientific™ Savant™ DNA SpeedVac™ Concentrators Rotors with Savant DNA SpeedVac Concentrators.
Easy to use circular rotors with different tube hole sizes to accommodate different sample sizes. These rotors require no additional accessories and fit directly into your concentrator.
Specifications
Description72 x 0.5mL microtubes
TypeConcentrator Rotor
For Use With (Equipment)DNA130
Holds72
Rotor SystemTraditional
Volume (Metric)0.5 mL
Unit SizeEA