Precision™ Low Temperature Refrigerated Incubator, 173 L
Actual product may vary
Precision™ Low Temperature Refrigerated Incubator, 173 L
Temperature Range: -10°C to +60°C
Have Questions?
Catalog NumberVoltage
PR205740R230 V
Catalog number PR205740R
HSN 84198990
Price (SGD)
8,215.00
Each
Voltage:
230 V
Price (SGD)
8,215.00
Each
Conduct applications that require temperatures ranging from -10° to +60°C with Thermo Scientific™ Precision™ Low Temperature BOD Refrigerated Incubators, available in 2 sizes and ideal for biochemical oxygen demand testing, bacterial research, seed germination, fish/insect breeding and much more. Features an efficient, dependable design with microprocessor controls and forced air circulation.
Temperature Uniformity
  • Microprocessor control with easy-to-read display shows actual temperature within 0.1°C
  • Push-button controls for temperature set point selection
  • Forced-air circulation delivers excellent temperature stability
  • CFC-free, foamed polyurethane insulation prevents heat loss
  • RTD temperature probe and protected set point mode prevent accidental temperature change
  • Door key lock protects samples from unauthorized access
Efficient Design
  • Easy-to-clean, corrosion-resistant construction
  • Compressor relay conserves energy
173L (6.1 cu. ft.) Unit
  • Economical undercounter design
  • High/low safety thermostat backups
  • Three cooling modes simplify operation: high precision with cooling; high precision without cooling; and frost-free with variable cooling
  • Four adjustable leveling feet for stable setup
  • 2A outlet easily supports apparatus inside the unit
566L (20 cu. ft.) Unit
  • Ideal for BOD applications and temperature settings at or below ambient
  • Holds over 300 BOD bottles (300mL)
  • Temperature set point selection with high- and low-temperature protection and simple calibration
  • Available with dual lamp fluorescent lighting for plant growth studies and day/night cycles, programmable lighting conditions
  • Safety relay and alarm LED alert to over/undertemperature conditions
  • Access port for independent sensors/connection of equipment inside unit
  • Cooling switch provides high temperature accuracy and saves energy at temperatures above ambient
  • RS-232 and recorder jacks for datalogging
  • Plant growth models available: PR505755L and PR505750L–with  Dual-lamp fluorescent lighting and programmable temperature and light cycles
849.5L (30.0 cu. ft.) Unit
  • Large volume for applications above ambient
  • Temperature range: 5° to 70°C
  • Hydraulic thermostat with analog temperature control
  • Choice of solid door or glass door for viewing of samples; no door lock
  • Easy-to-clean, corrosion- and chemical-resistant aluminum chamber
  • Requires hardwire installation by qualified electrician
Specifications
Capacity (English)6.1 cu. ft.
Capacity (Metric)173 L
Chamber MaterialAluminum
No. of Shelves2 std./4 max.
Temperature Range (Metric)-10°C to +60°C
TypeRefrigerated Incubator
Door StyleSolid
InsulationCFC-free, foamed polyurethane
Temperature Uniformity± 0.6/1.0°C @ 37°C/20°C
Dimensions (D x W x H) Interior20.5 x 20.5 x 28 in. (52 x 52 x 70 cm)
Dimensions (L x W x H)24.5 x 24 x 34.5 in. (62.2 x 61 x 87.6 cm)
Height (English)34.5 in.
Height (Metric)87.6 cm
Electrical Requirements230 V, 50 Hz, 5.8 A, Nema 6-15P plug
Voltage230 V
Amperage5.8 A
Frequency50 Hz
Unit SizeEach