MB90 Moisture Analyzer
MB90 Moisture Analyzer
Actual product may vary
Ohaus
MB90 Moisture Analyzer
Get both speed and precision in one moisture analyzer. The OHAUS™ MB90 uses halogen heating which distributes heat uniformly over the sample and speeds up the drying process, delivering fast and precise measurements. Designed for tool-free cleaning, the components can be easily removed for quick and easy maintenance. Operation is intuitive with the icon-driven menu navigation on the touchscreen display.
Have Questions?
Catalog number OHAMB90
View Price:Sign in to see your account pricing. Need an account? Register with us today.
  • ABS top housing, stainless steel pan support, in-use cover
  • Storage up to 2 methods and 100 test results for each method
  • User-guide, 13 operational languages
  • Precisely controlled halogen heating dries samples quickly and ensures uniform heating to yield fast, precise and repeatable results of 0.01% readability
  • Effortless Maintenance: removable pan handler and heating chamber components that make cleaning quick and easy, with no tools needed
  • Icon-based menu navigation on the easy-to-read touchscreen display makes operation intuitive
  • Perform routine sample testing in three easy steps; no training required
  • Two drying profiles: Customize heating of specific samples with temperature control for one-step drying
  • Customize your own test endpoint criteria to automatically end the test once moisture loss is no longer detected
  • Wide, bright touchscreen display and intuitive menu navigation for fast, accurate moisture analysis
  • Versatile connectivity options to handle data collection and transfer needs; USB and RS232 Ports; Data collection and transfer
Specifications
ModelMB90
OutputsRS232, USB device, USB host
Repeatability0.18% (3g sample), 0.02% (10g sample)
Certifications/ComplianceIEC/EN 61010-1; IEC/EN 61010-2-010; CAN/CSA C22.2
Dimensions (L x W x H)8.3 x 14.0 x 7.7 in. (211 x 355 x 180 mm)
Electrical Requirements120/240 VAC 50/60 Hz
Readability0.01%/0.001 g
Display TypeDigital LCD
Capacity90 g
Humidity RangeMoisture Range: 0.01% to 100% (0.01% to 1000% for
Net Weight (English)11.5 lb.
Sample Size0.5 g to 90 g
Temperature Range (Metric)10°C to 40°C
TypeMoisture Balances
Voltage120 V
Unit SizeEA