Tie-on Facemask with Visor
Actual product may vary
Tie-on Facemask with Visor
Optically clear, anti-fog visor combined with a tie-on, latex-free, low linting, bacterial filtering facemask. Nitritex™ Tie-on Facemask with Visor provides superior filtration efficiency, comfort, breathability and a visor for additional protection.
Have Questions?
Catalog number NRTVFM210-T-SLOT
Price (SGD)
552.00
Pack
Price (SGD)
552.00
Pack
  • Latex free
  • Low linting mask
  • High bacterial and particle filtration efficiency
  • Optically clear visor
  • Distortion free visor
  • Anti-fog visor
  • Tie-on
  • ISO Class 4 and EU GMP Grade A compatible
  • Plastic-coated steel noseband
Specifications
Product TypeFace Mask with Visor
ColorWhite
SterilitySterile
StylePleated with Ties
Certifications/ComplianceISO 9001:2008, ISO 2859
Construction3-ply
Dimensions (L x W x H)210 x 95 mm
Filter Efficiency>96%
IncludesVisor Facemask
Length (Metric)210 mm
ModelVFM201
Packaging Quantity100/Bag
Width (Metric)95 mm
Unit SizePack