BioClean™ Chem Prep Sterile Mat S-BCPM
Actual product may vary
Ansell
BioClean™ Chem Prep Sterile Mat S-BCPM
Sterile three-layered preparation mat features leak-proof properties and superb absorption.
Have Questions?
Catalog NumberDimensions (L x W)
NRTS-BCPM-5640B56 x 40 cm
Catalog number NRTS-BCPM-5640B
Price (SGD)
-
Dimensions (L x W):
56 x 40 cm
  • Three-layered construction: For highly leak-proof absorbency
  • Gamma irradiation sterilization: Suitable for use in an ISO 5 cleanroom
  • Manufactured in an ISO 9001 facility: For product quality assurance
N/A
Specifications
AbsorbancyHighly absorbant
Certifications/ComplianceManufactured in a facility holding ISO 9001:2008 certifications; Processed in an NEBB certified ISO Class 5 environment
Dimensions (L x W)56 x 40 cm
DisposableSingle-use
For Use With (Application)Controlled and Critical, Environments, Laboratory and Research, Production and Manufacturing
QuantityOne piece per sealed inner PE bag; one inner bag per second PE bag; 25 double bags per sealed outer PE bag; four outer bags per lined carton (100 pieces)
ColorBlue
Length (English)22 in.
Length (Metric)56 cm
MaterialPolyolefin
TypeAdhesive Floor Mat
Width (English)16 in
Width (Metric)40 cm
Unit SizeCase