BioClean ISO Leaf™ Cleanroom Bond Paper
BioClean ISO Leaf™ Cleanroom Bond Paper
Actual product may vary
BioClean ISO Leaf™ Cleanroom Bond Paper
Provides latex-free performance in critical environments. Nitritex™ BioClean ISO Leaf™ Cleanroom Bond Paper is heat resistant and ISO Class 4 compatible. It is available in 5 colors.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColor
NRTBICP-A4Y80Yellow
NRTBICP-A4B80Blue
NRTBICP-A4G80Green
NRTBICP-A4P80Pink
Catalog number NRTBICP-A4Y80
Price (SGD)
31.00
EA
Add to cart
Color:
Yellow
Price (SGD)
31.00
EA
Add to cart
  • Provides sharp, clear reproductions
  • Latex-free and heat-resistant
  • Processed to ensure ISO Class 4 compatibility
  • Manufactured in ISO 9001:2008 registered facilities
  • Lot number included on bag and carton to provide traceability
  • Store in a dry, cool place away from direct sunlight
  • Ideal storage range: 20-70% humidity, 10-40°C
Specifications
RuledNo
ColorYellow
Dimensions (L x W)297 x 210 mm
Length (Metric)297 mm
Width (Metric)210 mm
Certifications/ComplianceISO 9001:2015
Plain paperYes
Product TypeCleanroom Bond Paper
Quantity250 Sheets/Sealed PE Pk.
SizeA4
Weight (Metric)80 gsm, ± 3 g
Unit SizeEA